Orion Star™ A326 Multi-Parameter Meter (pH/RDO/DO), Handheld
Product Details & Documents
This rugged, waterproof portable meter is designed for a wide range of pH, mV, ORP, dissolved oxygen and temperature testing and field applications. It can be used in the most demanding locations thanks to its IP67-rated housing. The meter offers two measuring channels allowing users to measure pH and dissolved oxygen simultaneously or view each channel separately.
- Up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups
- Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
- Calibration log stores ten most recent calibrations
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
Measure pH, mV, relative mV or ORP with temperature on channel one and measure dissolved oxygen in percent saturation or mg/L with temperature using RDO optical DO probes or polarographic DO probes on channel two. Warrenty; 3 years.
Certifications: CE, TUV 3-1, FCC Class A.
Delivery information: Supplied with four AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate.
The kit additionally includes: 8107UWMMD ROSS gel-filled epoxy pH/ATC Triode (3 m cable), 087010MD RDO optical DO probe (3 m cable), pH 4,01/pH 7,00/pH 10,01 buffer pouches, Orion ROSS electrode storage solution, Orion 1413 µS conductivity standard pouches, rinse solution pouches, protective armor with electrode holder and hard carrying case.
Specifications
- Description:Handheld pH/DO meter Orion Star A326, instrument alone
- Seq:01
- Connections:BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature)
- Battery type:4 AA (included)
- Battery life:800 hours
- Compliance:IP 67
- DO concentration resolution:0,1 / 0,01 mg/L
- Memory:Non-volatile memory preserves data log, calibration log and meter settings
- IP rating:IP 67
- DO accuracy:Polarographic: ±0,2 mg/L or ±2% of reading, whichever is greater
RDO: ±0,1 mg/L (up to 8 mg/L), ±0,2 mg/L (8 - 20 mg/L), ±10% of reading (up to 50 mg/L) - Data logging capacity:5000 with date and time stamp
- Languages:English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
- Power supply:AC adapter or 4 AA batteries
- mV resolution:0,1
- Pressure accuracy:6 mm Hg
- DO calibration:Water-saturated air, air saturated water, manual (Winkler) and zero point
- Pressure resolution:1 mm Hg
- W×D×H:240×100×65 mm
- Temperature range:–5,0...+105,0 °C
- Temperature calibration:1 point temperature offset calibration
- mV range:±2000,0
- DO barometric pressure range:Automatic or manual, 450 - 850 mmHg
- No. of channels:2
- pH range:–2,000...+20,000
- Calibration:pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
- Temperature compensation:Automatic or manual
- pH calibration:Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
- Temperature resolution:0,1 °C, 0,1 °F
- DO concentration:0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
- Temperature accuracy:±0,1 °C
- Weight:0,5 kg
- Cat. No.:665-0279
- pH accuracy:±0,002
- pH resolution:0,1 / 0,01 / 0,001
- DO resolution:0.1, 1 % saturation
- Data logging - log function:Manual / Automatic with AUTO-READ™
- Alarm:High/low limit alarm, calibration due alarm
- Model:A326
- Outputs:RS232, USB
- mV accuracy:±0,2 mV or 0,05% of reading whichever is greater
- Calibration points:1 point relative mV (RmV) mode, 1 point ORP mode
- DO saturation:0,0 - 600,0 % (polarographic); 0,0 - 500,0 % (RDO)
- Display:Graphic LCD with backlight
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cablePrinter and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cable662-1140Thermo Scientific™ Orion™ Versa Star meters, Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus metersLog in for availability1 * 1 SET - 76,40 €
Product Information
- Catalog #AvailabilityPrice
- Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series metersSpare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series meters662-1547Log in for availability1 * 1 items - 72,40 €
Product Information
- Catalog #AvailabilityPrice
- Protective armor for portable meters, incl. hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, conductivity and dissolved oxygen sensor holderProtective armor for portable meters, incl. hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, conductivity and dissolved oxygen sensor holderORIOSTARA-ARLog in for availability1 * 1 items - 102,00 €
Product Information
- Catalog #AvailabilityPrice
- Orion™ Star A Series portable meter conductivity and DO probe holder, can be attached to either side of the portable meter armor and holds 12/15 mm Ø conductivity probes and a dissolved oxygen probe when placed in its calibration sleeveOrion™ Star A Series portable meter conductivity and DO probe holder, can be attached to either side of the portable meter armor and holds 12/15 mm Ø conductivity probes and a dissolved oxygen probe when placed in its calibration sleeveORIOSTARA-ESCDLog in for availability1 * 1 items - 29,50 €
Product Information
- Catalog #AvailabilityPrice
- Orion™ Star™ A Series portable meter pH electrode holder, can be attached to the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeveOrion™ Star™ A Series portable meter pH electrode holder, can be attached to the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeveORIOSTARA-ESPHLog in for availability1 * 1 items - 29,50 €
Product Information
- Catalog #AvailabilityPrice
- Hard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeveHard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeveORIOSTARA-CSLog in for availability1 * 1 items - 144,00 €
Product Information
- Catalog #AvailabilityPrice
- Soft-sided field case, incl. interior dividers and exterior pockets to safely organise the meter and accessories in the fieldSoft-sided field case, incl. interior dividers and exterior pockets to safely organise the meter and accessories in the fieldORIO1210005Log in for availability1 * 1 items - 161,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Cable adapter, for connecting the RS232 port of a meter to a USB portCable adapter, for connecting the RS232 port of a meter to a USB portORIO1010005Orion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series) and Orion™ Star and Star Plus metersLog in for availability1 * 1 items - 131,00 €
Product Information
- Catalog #ForAvailabilityPrice
- RS232 cable, for connecting a meter to a PC or printerRS232 cable, for connecting a meter to a PC or printerORIO1010053Orion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series), Orion™ Dual Star benchtop meters, Orion 3-Star, 4-Star and 5-Star metersLog in for availability1 * 1 items - 45,40 €



