Toluene ≥98%, TECHNICAL
Metal drum, 200 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Methylbenzene, Phenylmethane, FM approval
- CAS Number:108-88-3
- Formula:C₆H₅CH₃
- MDL Number:MFCD00008512
- Molecular Weight:92.14 g/mol
- Merck Index:13,09607
- Boiling Pt:110.6 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:0.867 g/cm³ (20 °C)
- Flash Pt:4 °C
- Storage Temperature:Ambient
- EINECS:203-625-9
- UN:1294
- ADR:3,II
- REACH:01-2119471310-51
Certificates
Search by Lot Number30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
P20_VWRC28701.364_SPEC_EN.pdf P20_VWRC28701.466_SPEC_EN.pdf P20_VWRC28701.550_SPEC_EN.pdf spec_VWRC28701.364 spec_VWRC28701.466
More
Specifications
Specification Test Results
- Assay:Min. 98 %
Specifications
- Pack type:Metal drum
- Pk:200 L



