0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant protein, d8s2298e( np_005662, 173 a.a. - 268 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
ABNOH00007993-Q01
:
Abnova
Recombinant protein, d8s2298e( np_005662, 173 a.a. - 268 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
ABNOH00007993-Q01
:
Abnova
sds_ABNOH00007993-Q01.pdf
TDS_ABNOH00007993-Q01.pdf