Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant protein, kit (aah71593.1) recombinant protein without tag. 1 * 10 µG
Catalog # ABNOH00003815-G01
Supplier:
Abnova
Expand image
Recombinant protein, kit (aah71593.1) recombinant protein without tag. 1 * 10 µG
Catalog # ABNOH00003815-G01
Supplier:
Abnova
Related Information
Documentation
Datasheet
TDS_ABNOH00003815-G01.pdf
Documentation