Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER SOLN 4+7+10+STRIPS 1 * 1 SET
32044292BOM_SG
:
VWR Chemicals
BUFFER SOLN 4+7+10+STRIPS 1 * 1 SET
32044292BOM_SG
:
VWR Chemicals
sds_VWRC32044