Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER SOLN 4+7+10+ STRIPS 1 * 1 SET
Catalog # 32095297BOM_SG
Supplier:
VWR Chemicals
SDS
Expand image
BUFFER SOLN 4+7+10+ STRIPS 1 * 1 SET
Catalog # 32095297BOM_SG
Supplier:
VWR Chemicals
SDS
Related Information
Documentation
SDS
sds_VWRC32095
Documentation