ContactUsLinkComponent
Recombinant protein, cryba4( np_001877.1, 1 a.a. - 196 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001413-P01
Supplier:  Abnova
Recombinant protein, cryba4( np_001877.1, 1 a.a. - 196 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001413-P01
Supplier:  Abnova
Related Information