0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant protein, crh( np_000747.1, 1 a.a. - 196 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
ABNOH00001392-P01
:
Abnova
Recombinant protein, crh( np_000747.1, 1 a.a. - 196 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
ABNOH00001392-P01
:
Abnova
sds_ABNOH00001392-P01.pdf
TDS_ABNOH00001392-P01.pdf