0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant protein, flot2( aah17292.1, 1 a.a. - 379 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00002319-P02
:
Abnova
Recombinant protein, flot2( aah17292.1, 1 a.a. - 379 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00002319-P02
:
Abnova
sds_ABNOH00002319-P02.pdf
TDS_ABNOH00002319-P02.pdf