QUANTOFIX® Test Strips, Chromate
QUANTOFIX® chlorine sensitive, 0 • 0,1 • 0,5 • 1 • 3 • 10 - mg/L - Cl₂; 0,1–10 - mg/L - Cl₂, Redox reaction (potassium iodide/redox indicator), 43×123×40 mm (packaging), 100 Tests, 100
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of chromate in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within 30 seconds one gets a reliable result.
Documents
Certificates
Search by Lot NumberDip and Read Tests, Rapid Tests.pdf
Additional Product Details
IFU QUANTOFIX® Chlor 500.pdf IFU Test strips, chlorine Sesitive, QUANTOFIX®.pdf IFU Test strips, chlorine, QUANTOFIX®.pdf
MANA91354_TDS_EN
Datasheet
Specifications
Specifications
- Chemical method:Redox reaction (potassium iodide/redox indicator)
- Pk:100
- Description:QUANTOFIX® chlorine sensitive
- Dimensions:43×123×40 mm (packaging)
- Tests per kit:100 Tests
- Range:0 • 0,1 • 0,5 • 1 • 3 • 10 - mg/L - Cl₂; 0,1–10 - mg/L - Cl₂
- Cat. No.:705-1156
- Parameters:All reagents included
- Contents:100 test strips in a tube, instruction leaflet
- Platform:Semi-quantitative test strips
- Compliance:No hazardous material
- Instrument:QUANTOFIX Relax
- Shelf life (months):30 (from production)
- Method:Chlorine - Redox reaction (yellow → violet)
- Weight:32 g



