ContactUsLinkComponent
PUR Container - Nomadic Pre-qualified 48h shipper - External 521 x 470 x 584 mm, Internal 254 x 203 x 203mm 1 * 1 items
Catalog # SCATE280CUPS
Supplier:  Sonoco Thermosafe
PUR Container - Nomadic Pre-qualified 48h shipper - External 521 x 470 x 584 mm, Internal 254 x 203 x 203mm 1 * 1 items
Catalog # SCATE280CUPS
Supplier:  Sonoco Thermosafe