Order Entry
Export
Orders LinkContactUsLinkComponent
illustra™ ExoProStar™ kits

illustra™ ExoProStar™ kits

illustra™ ExoProStar™ Enzymatic PCR and Sequence Reaction Clean Up Kit, 2.000 Tests
Catalog # US78212
Supplier:   Cytiva
illustra™ ExoProStar™ kits
Catalog # US78212
Supplier:  Cytiva
CAS Number:  
illustra™ ExoProStar™ kits
illustra™ ExoProStar™ kits

illustra™ ExoProStar™ Enzymatic PCR and Sequence Reaction Clean Up Kit, 2.000 Tests

CAS Number:  
Price
1 320,00 €
Log in for availability
1 * 2.000 Tests
Product Details & Documents

illustra™ ExoProStar™ contains illustra™ Alkaline Phosphatase and Exonuclease 1, formulated to work together to remove unincorporated primers and nucleotides from amplification reactions in preparation for sequencing, cloning, genotyping or further DNA modification reactions.

  • Enzymes optimised to work together for high efficiency removal of unincorporated primers and nucleotides
  • Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction
  • Fast 30 minute protocol; complete heat inactivation of the enzymes within 15 minutes
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate
Specifications

Specifications

  • Pk:
    2.000 Tests
  • Description:
    illustra™ ExoProStar™ Enzymatic PCR and Sequence Reaction Clean Up Kit
  • Cat. No.:
    US78212
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Target molecule:
    DNA
  • Method / Format:
    Enzymatic clean-up
  • Description:
    illustra™ ExoProStar™ Enzymatic PCR and Sequence Reaction Clean Up Kit
  • Sample type:
    PCR and other enzymatic reactions

Specifications