Orders LinkContactUsLinkComponent
Reagents for comparators
Reagents for comparators
DPD 1 (free chlorine), 250 tests, DPD, 250 Tab.
Catalog # PALIAT010
Supplier:   Palintest
Reagents for comparators
Catalog # PALIAT010
Supplier:  Palintest
CAS Number:  
Reagents for comparators
Reagents for comparators
DPD 1 (free chlorine), 250 tests, DPD, 250 Tab.
CAS Number:  
Price
52,50 €
Log in for availability
1 * 250 Tablet
Product Details & Documents
Comparator tablet reagents are designed for use with the contour comparator, standard comparator and other visual test platforms.
Specifications
Specifications
  • Chemical method:
    DPD
  • Pk:
    250 Tab.
  • Description:
    DPD 1 (free chlorine), 250 tests
Specifications