Flasks, round bottom, Quickfit® ground joints
1000 ml, 19/26 NS, 177 mm
Catalog # BIBBFR1L/2S
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Certifications: ISO 1773, ISO 4797.
Documents
Specifications
- Socket size:19/26 NS
- Capacity:1000 ml
- Height:177 mm
- Seq:30



