Order Entry
Export
Orders LinkContactUsLinkComponent
Accessories for DOSE IT

Accessories for DOSE IT

Silicone tubing, autoclavable, length: 2,5 m, 2 mm
Catalog # 710-1024
Supplier:   INTEGRA Biosciences
Accessories for DOSE IT
Catalog # 710-1024
Accessories for DOSE IT
Accessories for DOSE IT

Silicone tubing, autoclavable, length: 2,5 m, 2 mm

Price
33,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Documents

Specifications
  • Description:
    Silicone tubing, autoclavable, length: 2,5 m
  • Ø int.:
    2 mm