Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
VISCOSITY STANDARD N10
VISCOSITY STANDARD N10
Print
VISCOSITY STANDARD N10
Catalog # CANN9727-C32
Supplier:
CANNON INSTRUMENT
VISCOSITY STANDARD N10
Catalog # CANN9727-C32
Supplier:
CANNON INSTRUMENT
Expand image
Return to Top
VISCOSITY STANDARD N10
Add to cart
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 500 mL
-
+
Inquire For Price
Return Policy
Product Details & Documents