Orders LinkContactUsLinkComponent
Office desks
Office desks
Corner bracket, 800x800 mm, for linking 2 desks, quadrant, including 1 cable clamp
Catalog # HAMBVHE91/5
Supplier:   HAMMERBACHER
Office desks
Catalog # HAMBVHE91/5
Supplier:  HAMMERBACHER
Office desks
Office desks
Corner bracket, 800x800 mm, for linking 2 desks, quadrant, including 1 cable clamp
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Desk series with a range of variation and combination options.
Tabletop: 25 mm thick, melamine resin-coated, scratch-resistant panel in accordance with DIN 68765, with 2 mm rounded plastic edging.
Frame: Stove-enamelled, silver effect (RAL 9006), all around steel edging 20×40 mm; when linking 'straight' desks, the table foot can be placed at the back. Tubular table feet, 60 mm diameter.
Electrification: Horizontally with CKHO cable conduit, vertically with CKVE cable spiral (both available to order).
Height adjustment: 68 to 76 cm using levelling foot. Also adjustable to 88 cm with additional HHOE special height adjuster (available to order).
Delivery information: Easy to construct thanks to clear, comprehensible assembly instructions that are straightforward and quick to complete, even for novices.
Specifications
  • Description:
    Corner bracket, 800x800 mm, for linking 2 desks, quadrant, including 1 cable clamp
  • Seq:
    41