Cabinets with roll front
Product Details & Documents
Carcass construction, i.e. each cabinet can stand alone. Carcass firmly glued and assembled.
- 5 filing shelves can be used
- 8 mm visible rear panel
- With shelving
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Panels/base: 22 mm thick, with 2 mm ABS edging on the front. Height adjustable in 32 mm increments with recessed shelf supports. Sides and fronts 19 mm thick.
Roller shutters: High quality, PVC-free, one-piece, silver-coloured roller shutters, glide on tracks, not lockable. Roller shutters can remain open in any position.
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, cabinet can be made 5,1 cm higher with the steel base than with the adjustable feet.
Grip shape: Extruded and recessed handles
: The cabinets are delivered fully assembled.
Specifications
- Description:Cabinets with roll front, maple decor body, silver decor front
- Seq:03
Complementary Products
Product Information
- Catalog #DescriptionAvailabilityPrice



