Orders LinkContactUsLinkComponent
Supports for stand grids
Supports for stand grids
Flat flanges LF 200, framework grid 0,707 m
Catalog # LENZ06247521
Supplier:   Lenz Laborglas
Supports for stand grids
Catalog # LENZ06247521
Supplier:  Lenz Laborglas
Supports for stand grids
Supports for stand grids
Flat flanges LF 200, framework grid 0,707 m
Price
2 060,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel (clamp), steel, powder coated (supports).
For fixing lids onto reaction vessels and for the suspension into stands, complete with clamp and supports.
Specifications
  • For:
    Flat flanges LF 200, framework grid 0,707 m
  • Seq:
    04