Orders LinkContactUsLinkComponent
Fittings
Fittings
Luer adaptor, to connect 1/16" OD semi rigid tubing
Catalog # OMNI009LA
Supplier:   OMNIFIT
Fittings
Catalog # OMNI009LA
Supplier:  OMNIFIT
Fittings
Fittings
Luer adaptor, to connect 1/16" OD semi rigid tubing
Price
58,20 €
Log in for availability
1 * 5 items
Product Details & Documents
Documents
Specifications
  • Description:
    Luer adaptor, to connect 1/16" OD semi rigid tubing
  • Seq:
    68