Inlet liners
Cup splitter, high & low MW compounds, 5,0 mm, 3,5 mm, 100 mm
Catalog # REST20740
Supplier: Restek
Jump to:
Product Details & Documents
These liners can be used with Agilent GCs.
Ø int. = Nominal diameter at syringe needle expulsion point
Specifications
- Ext.Ø:5,0 mm
- Ø int.:3,5 mm
- Description:Cup splitter, high & low MW compounds
- Length:100 mm
- Seq:088



