Order Entry
Export
Orders LinkContactUsLinkComponent
 

Inlet liners

Baffle splitter, universal for the most common analyses, 6,2 mm, 1,0 mm, 92,1 mm
  REST20737
 :   Restek
Inlet liners
  REST20737
 :  Restek
Inlet liners
Inlet liners

Baffle splitter, universal for the most common analyses, 6,2 mm, 1,0 mm, 92,1 mm

 
 
   
1 * 5 items
 

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

 
  • Ext.Ø:
    6,2 mm
  • Ø int.:
    1,0 mm
  • Description:
    Baffle splitter, universal for the most common analyses
  • Length:
    92,1 mm
  • Seq:
    087