Order Entry
Export
Orders LinkContactUsLinkComponent
OX™ Cover Spectacles

OX™ Cover Spectacles

OX 3000: For use with earmuffs; anti UV (2C-1,2), anti-scratch, Blue, Clear PC
Catalog # 111-1633
Supplier:   3M
OX™ Cover Spectacles
Catalog # 111-1633
Supplier:  3M
OX™ Cover Spectacles
OX™ Cover Spectacles

OX 3000: For use with earmuffs; anti UV (2C-1,2), anti-scratch, Blue, Clear PC

Price
10,40 €
Log in for availability
1 * 1 items
Product Details & Documents

The 3M™ OX cover glasses (OTG) variants offer unique distinctive features: The OX2000 comes with adjustable lock-in temples (5 positions) and rotating tips (excellent grip in all positions) and the OX3000 is suitable for use with earmuffs; with low profile adjustable temples and non-slip pliable temple tips.

  • Optical Class 1 lens suitable for prolonged use
  • Protects against high speed particles at low energy (F) at extreme temperature conditions –5 and +55 °C (T)
  • Offers excellent protection against UV radiation
  • Designed to be worn over prescription eyewear

Can be used in a wide range of applications including construction, engineering, general assembly, light duty maintenance and repair and woodworking.

Certifications: EN 166:2001, EN 170:2002.

Specifications
  • Type:
    OX 3000: For use with earmuffs; anti UV (2C-1,2), anti-scratch
  • Colour:
    Blue
  • Lenses:
    Clear PC