1,3-Dioxane ≥98%
Glass bottle for solids, 25 g
: m-Dioxane, 1,3-Dioxacyclohexane
- :505-22-6
- :C₄H₈O₂
- :MFCD00006566
- :88.11 g/mol
- :105…106 °C
- :–45 °C
- :1.032
- :5 °C (41 °F)
- :Ambient
- :208-005-1
- :1165
- :3,II
Documents
- Pack type:Glass bottle for solids
- Pk:25 g