Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Dioxane ≥98%

Glass bottle for solids, 5 g
Catalog # H54107.06
Flammable
Irritant
1,3-Dioxane ≥98%
Catalog # H54107.06
CAS Number:  505-22-6
1,3-Dioxane ≥98%
1,3-Dioxane ≥98%

Glass bottle for solids, 5 g

CAS Number:  505-22-6
Price
70,50 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  m-Dioxane, 1,3-Dioxacyclohexane
  • CAS Number:505-22-6
  • Formula:C₄H₈O₂
  • MDL Number:MFCD00006566
  • Molecular Weight:88.11 g/mol
  • Boiling Pt:105…106 °C
  • Melting Pt:–45 °C
  • Density:1.032
  • Flash Pt:5 °C (41 °F)
  • Storage Temperature:Ambient
  • EINECS:208-005-1
  • UN:1165
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications