Sodium alginate
25 kg
Catalog # A3249.9025
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: Alginic acid sodium salt, Algin
- CAS Number:9005-38-3
- MDL Number:MFCD00081310
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:25 kg



