Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dimethyluracil

1,3-Dimethyluracil

Bottle, 25 g
Catalog # APOSOR27441-25G
Irritant
Supplier:   Apollo Scientific
1,3-Dimethyluracil
Catalog # APOSOR27441-25G
Supplier:  Apollo Scientific
CAS Number:  874-14-6
1,3-Dimethyluracil

Bottle, 25 g

CAS Number:  874-14-6
Price
24,60 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1,3-Dimethylpyrimidine-2,4(1H,3H)-dione
  • CAS Number:874-14-6
  • Formula:C₆H₈N₂O₂
  • MDL Number:MFCD00038065
  • Molecular Weight:140.14 g/mol
  • Melting Pt:119…125 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications