BOD measurement
Product Details & Documents
Measurement system without mercury for determination of the amount of dissolved oxygen consumed by aerobic biological microorganisms in water. The sensor uses manometric technology and micrprocessor control to provide regular data measurements over defined periods.
- Very simple to use and mercury-free
- Five values every 24 hours can be memorised or viewed instantaneously on the numerical display
- 1, 6 and 10 sample bottle versions available
- Measurement ranges: 90/250/600/999 ppm BOD
Temperature control is usually required for reproducible results - please see entries for thermostatic cabinets and incubators which are supplied separately.
Temperature control is usually required for reproducible results - please see entries for thermostatic cabinets and incubators which are supplied separately.
Delivery information: The BOD systems includes sensors, dark glass sample bottles, alkali holder for absorption of CO2, stirrer platform (not for single bottle vesrion) and stirrer bars.
Documents
Manual BOD Sensor.pdf
Manual
Specifications
- Description:BOD Sensor Set for a single sample analysis
Accessories
Product Information
- Catalog #AvailabilityPrice
- Dark glass bottle 500 ml for BOD systems
Product Information
- Catalog #AvailabilityPrice
- Control test tablets
Product Information
- Catalog #AvailabilityPrice
- Potassium hydroxide trap for BOD sensor