Order Entry
Export
Orders LinkContactUsLinkComponent
Recombinant streptavidin

Recombinant streptavidin

Streptavidin, recombinant, 5 mg
Catalog # 786-584
Supplier:   G-Biosciences
Recombinant streptavidin
Catalog # 786-584
Supplier:  G-Biosciences
CAS Number:  
Recombinant streptavidin
Recombinant streptavidin

Streptavidin, recombinant, 5 mg

CAS Number:  
Price
201,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Recombinant streptavidin from Streptomyces avidinii, produced in E.coli.

Streptavidin is a tetrameric protein that has no carbohydrate and thereby reduces the amount of non specific binding. The solubility of streptavidin (isoelectric pH 5) in aqueous buffer is lower than avidin, but the binding of streptavidin to biotin is similar to that of avidin.

Documents

Specifications

Specifications

  • Pk:
    5 mg
  • Description:
    Streptavidin, recombinant

Specifications