Order Entry
Export
Orders LinkContactUsLinkComponent
PIG® Poly utility trays

PIG® Poly utility trays

Poly utility trays, 2 L, 270×380×25 mm
Catalog # NEWPPAK656
Supplier:   NEW PIG
PIG® Poly utility trays
Catalog # NEWPPAK656
Supplier:  NEW PIG
PIG® Poly utility trays
PIG® Poly utility trays

Poly utility trays, 2 L, 270×380×25 mm

Price
21,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Flat-bottom poly trays are the economical way to store small parts. Great for lining your benches and shelving..

  • Light-coloured, flat-bottom design makes it easy to see screws, springs and other small parts
  • All-poly construction resists most chemicals; will not rust or corrode
  • Rotationally molded spill tray is ideal for applications that would compromise injected or thermoformed trays
  • Made from FDA-compliant resins for containment of food and cosmetic products

Square modular design allows efficient use of space on bench or shelves for process parts.

Specifications
  • Capacity:
    2 L
  • Description:
    Poly utility trays
  • Seq:
    01
  • W×D×H:
    270×380×25 mm