Orders LinkContactUsLinkComponent
Soxhlet extractors
Soxhlet extractors
Cone 29/32, 500 ml, 60/46 NS
Catalog # WITG2.212.500
Supplier:   witeg Labortechnik
Soxhlet extractors
Catalog # WITG2.212.500
Supplier:  witeg Labortechnik
Soxhlet extractors
Soxhlet extractors
Cone 29/32, 500 ml, 60/46 NS
Price
145,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass. For extraction from solids with solvents.
  • Very good chemical resistance
  • Very good temperature resistance
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.
Specifications
  • Extraction cap.:
    500 ml
  • Socket size:
    60/46 NS
  • Description:
    Cone 29/32
  • Seq:
    01