Order Entry
Export
Orders LinkContactUsLinkComponent
 

Disposal Boxes for Broken Glasses

with max. loading weight of 5,5 kg, for lab bench, small, 200×200×250 mm
Catalog # ROTHKN69.1
Supplier:   Roth Carl
Disposal Boxes for Broken Glasses
Catalog # ROTHKN69.1
Supplier:  Roth Carl
Disposal Boxes for Broken Glasses

with max. loading weight of 5,5 kg, for lab bench, small, 200×200×250 mm

Price
Inquire For Price
Log in for availability
1 * 6 items
Product Details & Documents

Disposal boxes are made of cardboard, with 50 μm thick polyethylene liner bag, which prevents leakage of liquid residues.

  • Large flap in lid for easy filling
  • Flat folded for space-saving storage
  • Printed with eye-catching graphics in four languages (English, German, French, Spanish)

Ideal for safe collection and disposal of defective glassware.

Specifications
  • Description:
    with max. loading weight of 5,5 kg, for lab bench, small
  • Seq:
    01
  • W×D×H:
    200×200×250 mm