Orders LinkContactUsLinkComponent
Drawer storage, crystal multiroir M5000
Drawer storage, crystal multiroir M5000
Box with 2 small drawers, M5000 A, Ext: 355×185×101 mm, PC
Catalog # MULC11109
Supplier:   MULTIROIR
Drawer storage, crystal multiroir M5000
Catalog # MULC11109
Supplier:  MULTIROIR
Drawer storage, crystal multiroir M5000
Drawer storage, crystal multiroir M5000
Box with 2 small drawers, M5000 A, Ext: 355×185×101 mm, PC
Price
959,00 €
Log in for availability
1 * 12 items
Product Details & Documents
Crystal Multiroir M5000 storage boxes are modular, combinable and provided with a capacity of five litres.
  • Perfect transparency with possibility of autoclave sterilisation
  • High impact resistance with resistant to shocks
  • Available as a large drawer or 2 small drawers with the possibility to interchange all drawers
Specifications
  • Description:
    Box with 2 small drawers, M5000 A
  • Material:
    PC
  • Seq:
    05
  • W×D×H:
    Ext: 355×185×101 mm