ROTILABO WIDE-NECK CANS (500ML) Made of HDPE. Leakproof. With press-in plugs and screw cap. Ideal for storing powders or granulated substances. Large wide-neck opening enables easy filling and cleaning. ( neck inside 70MM ) ( 86x107MM ) 1 * 50 items
Catalog # ROTHHT58.1
ROTILABO WIDE-NECK CANS (500ML) Made of HDPE. Leakproof. With press-in plugs and screw cap. Ideal for storing powders or granulated substances. Large wide-neck opening enables easy filling and cleaning. ( neck inside 70MM ) ( 86x107MM ) 1 * 50 items
Catalog # ROTHHT58.1