Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Test tube, flat bottom, 16x50, 0.9mm, sodaglass. 1 * 200 items
Catalog # SCERA607016001021
Supplier:
DWK Life Sciences
Expand image
Test tube, flat bottom, 16x50, 0.9mm, sodaglass. 1 * 200 items
Catalog # SCERA607016001021
Supplier:
DWK Life Sciences