Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Storage cabinet, extension 100/60/200,blue 1 * 1 items
Catalog # SOVEC33907000
Supplier:
TRESTON
Expand image
Storage cabinet, extension 100/60/200,blue 1 * 1 items
Catalog # SOVEC33907000
Supplier:
TRESTON