0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Storage cabinet, extension 100/40/200 ,blue 1 * 1 items
SOVEC33707000
:
TRESTON
Storage cabinet, extension 100/40/200 ,blue 1 * 1 items
SOVEC33707000
:
TRESTON