0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Storage system 100/30/200 light grey,Standard part 1 * 1 items
Storage system 100/30/200 light grey,Standard part 1 * 1 items
Storage system 100/30/200 light grey,Standard part 1 * 1 items
SOVEC31335002
:
TRESTON
Storage system 100/30/200 light grey,Standard part 1 * 1 items
SOVEC31335002
:
TRESTON
Expand image
Storage system 100/30/200 light grey,Standard part 1 * 1 items
Add to cart
Unit of Measure
1 * 1 items
-
+