Order Entry
Export
Orders LinkContactUsLinkComponent
 

Test tubes, without rim, medium wall, `SAMCO 50`

100 mm, 16 mm, 1,2 mm
Catalog # MURRG023/34
Supplier:   MURRAY S
Test tubes, without rim, medium wall, `SAMCO 50`
Catalog # MURRG023/34
Supplier:  MURRAY S
Test tubes, without rim, medium wall, `SAMCO 50`
Test tubes, without rim, medium wall, `SAMCO 50`

100 mm, 16 mm, 1,2 mm

Price
81,10 €
Log in for availability
1 * 100 items
Product Details & Documents

Test tubes, made of SAMCO neutral glas.

Neutral glass test tubes will withstand moderate temperature change, they should not be taken from cold and placed directly into the hottest part of the flame without pre-heating.

Specifications
  • Ext.Ø:
    16 mm
  • Wall thickness:
    1,2 mm
  • Length:
    100 mm
  • Seq:
    04