Order Entry
Export
Orders LinkContactUsLinkComponent
MICRO SWITCH PB1 101 301 10A 250V 1 * 1 items

MICRO SWITCH PB1 101 301 10A 250V 1 * 1 items

Catalog # GERH1000819
Supplier:   GERHARDT
MICRO SWITCH PB1 101 301 10A 250V 1 * 1 items
Catalog # GERH1000819
Supplier:  GERHARDT
MICRO SWITCH PB1 101 301 10A 250V 1 * 1 items
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents