Shelving systems for drums, WSC-G-E
Drum storage system, with water protection compartment and 2 levels of shelves, with sump capacity 450 l, 12 - 20×200 l drums, 3130×1390×2230 mm
Catalog # LACOC22-5502-B
Supplier: LACONT UMWELTTECHNIK
Jump to:
Product Details & Documents
Water protection compartment containers are suitable for the storage of hazardous substances indoors.
Storage can take place both directly on the grid supports of the different levels and also on euro- or chemical pallets. The compact modular design allows an uncomplicated change of location in a very short time.
Specifications
- Capacity:12 - 20×200 l drums
- Description:Drum storage system, with water protection compartment and 2 levels of shelves, with sump capacity 450 l
- Seq:02
- W×D×H:3130×1390×2230 mm



