Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PE-AUFFANGWANNE, 805 X 640 X 335 MM 1 * 1 ST
Catalog # LACOP70-7046-A
Supplier:
LACONT UMWELTTECHNIK
Expand image
PE-AUFFANGWANNE, 805 X 640 X 335 MM 1 * 1 ST
Catalog # LACOP70-7046-A
Supplier:
LACONT UMWELTTECHNIK