Orders LinkContactUsLinkComponent
Sample cups for analysers, ratiolab®
For Technicon T1 analysers, 2 ml
Catalog # RATI5700110
Supplier:   RATIOLAB
Sample cups for analysers, ratiolab®
Catalog # RATI5700110
Supplier:  RATIOLAB
Sample cups for analysers, ratiolab®
Sample cups for analysers, ratiolab®
For Technicon T1 analysers, 2 ml
Price
17,70 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Made of clear PS.
  • Good chemical resistance
  • Good temperature resistance
Analysis beakers offer the highest precision for use with robotic analytical systems.
Specifications
  • Capacity:
    2 ml
  • Description:
    For Technicon T1 analysers
  • Seq:
    02