ContactUsLinkComponent
Precision thermometer, stem form ASTM 120 C, IP 92 C Kinematic Viscosity, Capillaries form: yellow enamelled round Hg filling range: +38,6+41,4C in 0.05C auxiliary scale: 0C, immersion length: total, total length: 300 mm, suitable for calibration 1 * 1 items
Catalog # SCNL1202120
undefined
Precision thermometer, stem form ASTM 120 C, IP 92 C Kinematic Viscosity, Capillaries form: yellow enamelled round Hg filling range: +38,6+41,4C in 0.05C auxiliary scale: 0C, immersion length: total, total length: 300 mm, suitable for calibration 1 * 1 items
Catalog # SCNL1202120