Bulb pipettes, Qualicolor, colour coded (one mark), Class B
50 ml, 0,100 ± ml, Red
KAVA632433131025
Soda-lime glass.
- Enlarged form
- Not autoclavable at 121 °C
: DIN 12690, ISO 648
- Colour code:Red
- Capacity:50 ml
- Seq:07
- Tolerance:0,100 ± ml
Soda-lime glass.
: DIN 12690, ISO 648