Order Entry
Export
Orders LinkContactUsLinkComponent
Bulb pipettes, Qualicolor, colour coded (one mark), Class B

Bulb pipettes, Qualicolor, colour coded (one mark), Class B

2 ml, 0,020 ± ml, Orange
Catalog # KAVAN632433131016
Supplier:   KAVALIER GLASS
Bulb pipettes, Qualicolor, colour coded (one mark), Class B
Catalog # KAVAN632433131016
Supplier:  KAVALIER GLASS
Bulb pipettes, Qualicolor, colour coded (one mark), Class B
Bulb pipettes, Qualicolor, colour coded (one mark), Class B

2 ml, 0,020 ± ml, Orange

Price
594,00 €
Log in for availability
1 * 50 items
Product Details & Documents

Soda-lime glass.

  • Enlarged form
  • Not autoclavable at 121 °C

Certifications: DIN 12690, ISO 648

Specifications
  • Colour code:
    Orange
  • Capacity:
    2 ml
  • Seq:
    02
  • Tolerance:
    0,020 ± ml