Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

50 ml, 0,050 ± ml, Red
Catalog # KAVA632433111025
Supplier:   KAVALIER GLASS
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Catalog # KAVA632433111025
Supplier:  KAVALIER GLASS
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

50 ml, 0,050 ± ml, Red

Price
26,70 €
Log in for availability
1 * 1 items
Product Details & Documents

Soda-lime glass.

  • Enlarged form
  • Non autoclavable at 121 °C

 : DIN 12691, ISO 648

Specifications
  • Colour code:
    Red
  • Capacity:
    50 ml
  • Seq:
    10
  • Tolerance:
    0,050 ± ml