7-Hydroxycoumarin, Sigma-Aldrich®
Glass bottle for solids, 25 g
SIAL93979-25G
: Merck
: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
- :93-35-6
- :C₉H₆O₃
- :MFCD00006878
- :162.14 g/mol
- :12,09981
- :382 °C (1013 hPa)
- :204…206 °C
- :Ambient
- :202-240-3
- Pack type:Glass bottle for solids
- Pk:25 g