Buffer, HEPES solution (1 mol/L), Sigma-Aldrich®
HEPES buffer solution (1 mol/l), Glass bottle, 500 mL
Catalog # SIAL83264-500ML-F
Supplier: Merck
Buffer, HEPES solution (1 mol/L), Sigma-Aldrich®
Catalog # SIAL83264-500ML-F
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:HEPES buffer solution (1 mol/l)



