Tween® 20 (Polysorbate), Sigma-Aldrich®
Glass bottle for solids, 1 kg
SIAL93773-1KG
: Merck
: Polyoxyethylene sorbitan monolaurate
- :9005-64-5
- :C₅₈H₁₁₄O₂₆
- :MFCD00165986
- :1227.53 g/mol
- :1.1 g/cm³ (20 °C)
- :Ambient
- :500-018-3
- Pack type:Glass bottle for solids
- Pk:1 kg